Learn More
Idex Threaded Plugs and Caps
Plugs to close off unused ports in valves and multi-port connectors
- Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port
Supplier: Idex UPP754
Description
Plugs to close off unused ports in valves and multi-port connectors
- Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port
Specifications
| Cap | |
| Yellow | |
| 5/16-24 | |
| Tefzel |