Learn More
Upchurch Scientific™ Nut
Supplier: Upchurch Scientific™ UP F 195X
Spécifications
| Nut | |
| 0.37 in. | |
| 125°C | |
| PEEK, Stainless Steel | |
| 1/16 in. O.D. Tubing |
| 0.64 in. | |
| 9000 psi | |
| Short Nuts, Standard Head, 10-32 | |
| Black |