Learn More
Thermo Scientific™ Universal Piping for Thermo Scientific™ 1300 Series A2 Biological Safety Cabinets
Factory-installed Universal Air/Gas Piping
$2,059.18 - $3,123.68
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|
13261354
|
Thermo Scientific™
1910288 |
5 ft. models, right and top/bottom |
Each for $3,123.68
|
|
|||||
|
13261352
|
Thermo Scientific™
1910286 |
ft. models, right and top/bottom |
Each for $2,565.27
|
|
|||||
|
13261355
|
Thermo Scientific™
1910289 |
5 ft. models, left and top/bottom |
Each for $3,123.68
|
|
|||||
|
13261403
|
Thermo Scientific™
1911418 |
4 ft. models, left and top |
Each for $2,059.18
|
|
|||||
|
13261405
|
Thermo Scientific™
1911420 |
5 ft. models, left and top |
Each for $2,382.93
|
|
|||||
Description
Universal Piping for Air/Gas Service
- Allows air and gas service to be piped through top, sides, or bottom of cabinet to minimize side-to-side footprint of cabinet utility piping
- Two pre-plumbed service ports per side only
- Factory-installed; must be ordered with cabinet
- For cabinets with stainless-steel liners, specify using unique part number rather than ordering left and right side separately
Order Info
Factory installed. Must be specified when ordering cabinet.