Learn More
Thermo Scientific™ Universal Piping for Thermo Scientific™ 1300 Series A2 Biological Safety Cabinets
Catalog No. 13261405
Change view
Click to view available options
For Use With (Equipment):
4 ft. models, left and top
5 ft. models, left and top
5 ft. models, left and top/bottom
5 ft. models, right and top/bottom
ft. models, right and top/bottom
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13261405 5 ft. models, left and top
13261354 5 ft. models, right and top/bottom
13261352 ft. models, right and top/bottom
13261355 5 ft. models, left and top/bottom
13261403 4 ft. models, left and top
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 13261405 Supplier Thermo Scientific™ Supplier No. 1911420
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Factory-installed Universal Air/Gas Piping
Universal Piping for Air/Gas Service
  • Allows air and gas service to be piped through top, sides, or bottom of cabinet to minimize side-to-side footprint of cabinet utility piping
  • Two pre-plumbed service ports per side only
  • Factory-installed; must be ordered with cabinet
  • For cabinets with stainless-steel liners, specify using unique part number rather than ordering left and right side separately
Order Info
Factory installed. Must be specified when ordering cabinet.
Specifications
Product Type Universal Piping
For Use With (Equipment) 5 ft. models, left and top