Learn More
Thermo Scientific™ Universal Piping for Thermo Scientific™ 1300 Series A2 Biological Safety Cabinets
Factory-installed Universal Air/Gas Piping
Supplier:  Thermo Scientific™ 1911418
 View more versions of this product
Catalog No. 13261403
Only null left
Description
Description
Universal Piping for Air/Gas Service
  • Allows air and gas service to be piped through top, sides, or bottom of cabinet to minimize side-to-side footprint of cabinet utility piping
  • Two pre-plumbed service ports per side only
  • Factory-installed; must be ordered with cabinet
  • For cabinets with stainless-steel liners, specify using unique part number rather than ordering left and right side separately
Order Info
Factory installed. Must be specified when ordering cabinet.
Specifications
Specifications
Universal Piping
4 ft. models, left and top
Product Suggestions
Product Suggestions