Learn More
Premier Lab Supply 32mm XRF Sample Cups PN# SC-4131
Catalog No. NC0043612
Encompass
32mm Single-Open Ended XRF Sample Cup with Cup, Ring & Collar (PN# SC-4131) Suitable for Universal OD: 32mm; ID: 24mm; Height: 23mm; Volume: 10mL 100 per bag
Catalog No. NC0043612 Supplier Premier Lab Supply Supplier No. SC4131
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
32mm Single-Open Ended XRF Sample Cup with Cup, Ring & Collar (PN# SC-4131) Suitable for Universal OD: 32mm; ID: 24mm; Height: 23mm; Volume: 10mL 100 per bag