Learn More
United Scientific Supplies United Scientific™ Contour Model Kit
Description
- Complete cartography kit for hands-on map creation
- Demonstrates contour line principles through water-level tracing
- Includes polystyrene landform model and precision container
- Accommodates 5-student groups with all materials provided
- Includes activity guide
Specifications
Specifications
| Type de produit | Cartography Kit |
| Comprend | Activity Guide |
| Grade | Intermediate - Middle School, High School, Higher Ed |