missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ UniStore Wide Mouth Square Bottles HDPE
UniStore Wide Mouth Square Bottles have are made of HDPE, are leakproof and have a polypropylene cap.
$36.25 - $93.36
Specifications
| Color | Translucent White |
|---|---|
| Material | HDPE |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
S01882
|
United Scientific™
33602 |
250 mL |
Pack of 12 Each for $74.79
|
|
|||||
|
S01884
|
United Scientific™
33604 |
1000 mL |
Pack of 6 Each for $93.36
|
|
|||||
|
S01886
|
United Scientific™
33601 |
175 mL |
Pack of 12 Each for $56.14
|
|
|||||
|
S01885
|
United Scientific™
33600 |
60 mL |
Pack of 12 Each for $36.25
|
|
|||||
|
S01883
|
United Scientific™
33603 |
500 mL |
Pack of 12 Each for $93.36
|
|
|||||
Description
UniStore™ Wide Mouth Square Bottles are available in different materials and sizes for multiple applications. The high-density polyethylene bottles (HDPE) have excellent chemical resistance to most acids, bases, and alcohols. The square design allows the bottles to nest together for more efficient storage, packing and shipping. These HDPE bottles are leakproof and have a polypropylene cap.
- Square design for compact storage
- Excellent chemical resistance
- Precision molded for consistent wall thickness with smooth interior and exterior surfaces
- FDA-21CFR compliant resin
- Leakproof closure
- Available in 60 mL, 175 mL, 250 mL, 500 mL, and 1000 mL
Specifications
| Translucent White | |
| Non-sterile | |
| No | |
| Reagent Bottle |
| HDPE | |
| Yes | |
| Wide, Threaded | |
| Not Autoclavable |