missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ UniStore Wide Mouth Square Bottles HDPE

Catalog No. S01882
Change view
Click to view available options
Capacity (Metric):
60 mL
175 mL
250 mL
500 mL
1000 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S01882 250 mL
S01884 1000 mL
S01886 175 mL
S01885 60 mL
S01883 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S01882 Supplier United Scientific™ Supplier No. 33602
Only null left

UniStore Wide Mouth Square Bottles have are made of HDPE, are leakproof and have a polypropylene cap.

UniStore™ Wide Mouth Square Bottles are available in different materials and sizes for multiple applications. The high-density polyethylene bottles (HDPE) have excellent chemical resistance to most acids, bases, and alcohols. The square design allows the bottles to nest together for more efficient storage, packing and shipping. These HDPE bottles are leakproof and have a polypropylene cap.

  • Square design for compact storage
  • Excellent chemical resistance
  • Precision molded for consistent wall thickness with smooth interior and exterior surfaces
  • FDA-21CFR compliant resin
  • Leakproof closure
  • Available in 60 mL, 175 mL, 250 mL, 500 mL, and 1000 mL

Specifications

Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Translucent White
Material HDPE
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide, Threaded
Product Type Reagent Bottle
Autoclavable Not Autoclavable