missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ UniStore Wide Mouth Square Bottles HDPE
Description
UniStore™ Wide Mouth Square Bottles are available in different materials and sizes for multiple applications. The high-density polyethylene bottles (HDPE) have excellent chemical resistance to most acids, bases, and alcohols. The square design allows the bottles to nest together for more efficient storage, packing and shipping. These HDPE bottles are leakproof and have a polypropylene cap.
- Square design for compact storage
- Excellent chemical resistance
- Precision molded for consistent wall thickness with smooth interior and exterior surfaces
- FDA-21CFR compliant resin
- Leakproof closure
- Available in 60 mL, 175 mL, 250 mL, 500 mL, and 1000 mL
Specifications
Specifications
| Capacity (Metric) | 175 mL |
| Color | Translucent White |
| Material | HDPE |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide, Threaded |
| Autoclavable | Not Autoclavable |
| Capacity (English) | 6 oz. |
| Product Type | Reagent Bottle |