missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ UniStore™ Graduated Wide Mouth Bottle
High quality translucent polypropylene with easy to read mL volume increments
$91.11 - $159.43
Specifications
| Color | Translucent |
|---|---|
| Material | Polypropylene |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | Yes |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
S01470
|
United Scientific™
33700 |
125 mL |
Pack of 12 Each for $91.11
|
|
|||||
|
S01471
|
United Scientific™
33701 |
250 mL |
Pack of 12 Each for $128.11
|
|
|||||
|
S01472
|
United Scientific™
33702 |
500 mL |
Pack of 12 Each for $159.43
|
|
|||||
|
S01473
|
United Scientific™
33703 |
1000 mL |
Pack of 6 Each for $121.00
|
|
|||||
Description
- These high-quality translucent polypropylene wide mouth bottles include easy to read mL volume increments
- The graduation lines are screen printed for sharp contrast, and each bottle has a white write-on area for identification
- Bottles are autoclavable and are ideal for storing liquids or solids
- Easy to fill wide mouth design
- Screen printed volume graduations for sharp contrast
- Write-on area for labeling
- Autoclavable at 121°C
- Leakproof
Specifications
| Translucent | |
| Non-sterile | |
| Yes | |
| Reagent Bottle |
| Polypropylene | |
| Yes | |
| Wide, Threaded | |
| Blank |