missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ UniStore™ Graduated Wide Mouth Bottle
Description
- These high-quality translucent polypropylene wide mouth bottles include easy to read mL volume increments
- The graduation lines are screen printed for sharp contrast, and each bottle has a white write-on area for identification
- Bottles are autoclavable and are ideal for storing liquids or solids
- Easy to fill wide mouth design
- Screen printed volume graduations for sharp contrast
- Write-on area for labeling
- Autoclavable at 121°C
- Leakproof
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Capacity (English) | 32 oz. |
| Color | Translucent |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | Yes |
| Mouth | Wide, Threaded |
| Product Type | Reagent Bottle |
| Chemical Label | Blank |
| Show More |