missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ UniStore™ Graduated Wide Mouth Bottle
High quality translucent polypropylene with easy to read mL volume increments
Supplier: United Scientific™ 33701
Description
- These high-quality translucent polypropylene wide mouth bottles include easy to read mL volume increments
- The graduation lines are screen printed for sharp contrast, and each bottle has a white write-on area for identification
- Bottles are autoclavable and are ideal for storing liquids or solids
- Easy to fill wide mouth design
- Screen printed volume graduations for sharp contrast
- Write-on area for labeling
- Autoclavable at 121°C
- Leakproof
Chemical Identifiers
| Translucent | |
| Non-sterile | |
| Yes | |
| Reagent Bottle |
| Polypropylene | |
| Yes | |
| Wide, Threaded | |
| Blank |
Specifications
| 250 mL | |
| Translucent | |
| Non-sterile | |
| Yes | |
| Reagent Bottle | |
| Autoclavable |
| 8 oz. | |
| Polypropylene | |
| Yes | |
| Wide, Threaded | |
| Blank |