Learn More
IDEX Health & Science LLC MicroTightUnions Assembly 1/32""
Numéro de catalogue. NC1477189
Encompass
Our MicroTight Unions Assembly is used specifically to easily connect capillary tubing together. True ZDV (Zero Dead Volume) allows two pieces of tubing to connect directly to each other using the included gauge and plug to ensure proper alignment. This allows for an uninterrupted fluidic path. It is for use with 1/32OD Tubing and includes (2) F-126S Fittings and (1) P-553 Gauge.
Numéro de catalogue. NC1477189 Fournisseur IDEX Health & Science LLC Code fournisseur P771
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Our MicroTight Unions Assembly is used specifically to easily connect capillary tubing together. True ZDV (Zero Dead Volume) allows two pieces of tubing to connect directly to each other using the included gauge and plug to ensure proper alignment. This allows for an uninterrupted fluidic path. It is for use with 1/32OD Tubing and includes (2) F-126S Fittings and (1) P-553 Gauge.