missing translation for 'onlineSavingsMsg'
Learn More

Undecane 99.5+%, TCI America™

Catalog No. S02835ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02835ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02835ML Supplier TCI America Supplier No. S02835ML

Chemical Identifiers

CAS 1120-21-4
Molecular Formula C11H24
Molecular Weight (g/mol) 156.31
MDL Number MFCD00008959
InChI Key RSJKGSCJYJTIGS-UHFFFAOYSA-N
Synonym n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und
PubChem CID 14257
ChEBI CHEBI:46342
IUPAC Name undecane
SMILES CCCCCCCCCCC

Specifications

Melting Point -26°C
Color Colorless
Boiling Point 195°C
Quantity 5 mL
UN Number 2330
Formula Weight 156.31
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Undecane [Standard Material for GC]
EINECSNumber (2)-0010
RTECSNumber YQ1525000
TSCA Yes