missing translation for 'onlineSavingsMsg'
Learn More

Sartorius ULTROSER G

Catalog No. 14558659
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14558659 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14558659 Supplier Sartorius Supplier No. 15950017

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ULTROSER G 20 mL serum substitute can successfully replace FCS (fetal calf serum) in cell culture medium.

  • Ultroser™ G serum substitute can successfully replace FCS (fetal calf serum) in cell culture medium
  • Designed for the in vitro culture of anchorage-dependent cells
  • Features amp; BenefitsSemi-defined composition ensuring batch-to-batch reproducibility
  • Low protein content
  • Easy use and storage
  • Ultroser G serum substitute has a concentration five times higher than fetal calf serum (2 % of reconstituted Ultroser G serum substitute is equivalent to 10% fetal calf serum in the basal medium)
  • Ultroser G is for research use only and not for use in diagnostic procedures

Specifications

Country of Origin United States
Form Liquid
Format Bottle
Product Type Serum Replacement
Quantity 20 mL
Sterility Sterile