Learn More
MP Biomedicals™ Ultra-Sense Femto-Plus Western ECL Substrate
Catalog No. MP08X100003
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
MP08X100003 100 mL
1 options
Catalog No. MP08X100003 Supplier MP Biomedicals™ Supplier No. 08X100003
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ultra-Sense Femto-Plus Western ECL Substrate has a high degree of sensitivity and enhanced chemiluminescence for femtogram detection of protein. It is optimized for use with PVDF and nitrocellulose membranes.
  • No protocol optimization required.
  • Easy-to-Use, just mix the solution A and B (1:1) for 30 seconds prior to use.
  • High degree of sensitivity and enhanced chemiluminescence for femtogram detection of protein.
  • Optimized for use with PVDF and nitrocellulose membranes.
  • Compatible with Western Blotting Markers.
  • Optimized for film- and CCD-based imaging.
Specifications
Quantity 100 mL
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.