Learn More
MP Biomedicals™ Ultra-Blue Fast PAGE Gel Staining Solution
Catalog No. MP08X100001
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
MP08X100001 1 L
1 options
Catalog No. MP08X100001 Supplier MP Biomedicals™ Supplier No. 08X100001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ultra-Blue Fast PAGE Gel Staining Solution next-generation formula offers a faster protein detection and higher sensitivity. The gels staining efficiency is innovatively enhanced by proprietary nanotechnology.
  • Staining efficiency is innovatively enhanced by proprietary nanotechnology.
  • Its next-generation formula offers a faster protein detection and higher sensitivity and there is no need for distaining.
  • No hazardous substances such as methanol and acetic acid, this gel staining solution is safe and environmentally friendly.
  • The product is also compatible with mass spectrophotometry.
Specifications
Quantity 1 L
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.