missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UEVLD Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561440
Description
Immunogen sequence: GKEVWVIGEQ GEDKVLTWSG QEEVVSHTSQ VQLSNRAMEL LRVKGQRSWS VGLSVADMVD SIVNNKKKVH SVSALA Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.
Possible negative regulator of polyubiquitination.Identifiants chimiques
| UEVLD | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| UEVLD | |
| 8430408E05Rik; Attp; EV and lactate/malate dehydrogenase domain-containing protein; signaling molecule ATTP; ubiquitin-conjugating enzyme E2 variant 3; UEV and lactate/malate dehyrogenase domains; Uev3; UEV-3; UEVLD | |
| UEVLD | |
| Primary | |
| Antigen affinity chromatography |
| 0.05 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IX04 | |
| 55293 | |
| Recombinant protein corresponding to Human UEVLD. Recombinant protein control fragment (Product #RP-98493). | |
| Antibody |
Spécifications
| UEVLD | |
| Polyclonal | |
| Unconjugated | |
| UEVLD | |
| 8430408E05Rik; Attp; EV and lactate/malate dehydrogenase domain-containing protein; signaling molecule ATTP; ubiquitin-conjugating enzyme E2 variant 3; UEV and lactate/malate dehyrogenase domains; Uev3; UEV-3; UEVLD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55293 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IX04 | |
| UEVLD | |
| Recombinant protein corresponding to Human UEVLD. Recombinant protein control fragment (Product #RP-98493). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |