missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ UDP-alpha-D-N-Acetylgalactosamine, Disodium Salt, >98% Calbiochem™,

Catalog No. 6701055MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6701055MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6701055MG Supplier MilliporeSigma™ Supplier No. 6701055MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Useful in synthesis of aryl azide derivatives that can be used in affinity labeling of glycosyltransferases and UDP-HexNAc pyrophosphorylase

Chemical Identifiers

CAS 108320-87-2
Molecular Formula C17H25N3Na2O17P2
Molecular Weight (g/mol) 651.32
MDL Number MFCD00077894
InChI Key HXWKMJZFIJNGES-UHFFFAOYNA-L
Synonym uridine 5'-diphospho-n-acetylgalactosamine disodium salt, disodium 2r,3s,4r,5r-5-2,4-dioxo-3h-pyrimidin-1-yl-3,4-dihydroxyoxolan-2-yl methyl 2r,3r,4r,5r,6r-3-acetamido-4,5-dihydroxy-6-hydroxymethyl oxan-2-yl phosphonato oxyphosphonate, uridine 5-diphospho-n-acetylgalactosamine disodium salt, 2-acetamido-2-deoxy-a-d-galactopyranosyl uridine diphosphate disodium salt, uridine 5' diphospho 1 2-acetamino-2-deoxy-alpha-d-galactopyranose disodium salt
PubChem CID 91864731
IUPAC Name disodium [5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxy}phosphonate
SMILES [Na+].[Na+].CC(=O)NC1C(O)C(O)C(CO)OC1OP([O-])(=O)OP([O-])(=O)OCC1OC(C(O)C1O)N1C=CC(=O)NC1=O

Specifications

Color White
Quantity 5 mg
Solubility Information Easily soluble in cold water
Formula Weight 651.3
Percent Purity ≥98%
Grade HPLC
Physical Form Solid
Chemical Name or Material UDP-alpha-D-N-Acetylgalactosamine, Disodium Salt
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above -20°C (-4°F).