missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ UDP-alpha-D-Galactose, Disodium Salt, >80% Calbiochem™,

Catalog No. 670120250MG
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
670120250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 670120250MG Supplier MilliporeSigma™ Supplier No. 670120250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 117756-22-6
Molecular Formula C15H22N2Na2O17P2
Molecular Weight (g/mol) 610.27
MDL Number MFCD00216890
InChI Key PKJQEQVCYGYYMM-UHFFFAOYNA-L
Synonym uridine 5'-diphosphoglucose disodium, disodium 2s,3r,4s,5s-5-2,4-dioxo-3h-pyrimidin-1-yl-3,4-dihydroxyoxolan-2-yl methyl 2s,3s,4r,5r,6s-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl phosphonato oxyphosphonate
PubChem CID 56846105
IUPAC Name disodium [5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxy}phosphonate
SMILES [Na+].[Na+].OCC1OC(OP([O-])(=O)OP([O-])(=O)OCC2OC(C(O)C2O)N2C=CC(=O)NC2=O)C(O)C(O)C1O

Specifications

Color White to Yellow
Quantity 250 mg
Solubility Information Easily soluble in cold water
Formula Weight 610.3
Percent Purity ≥80%
Physical Form Solid
Chemical Name or Material UDP-alpha-D-Galactose, Disodium Salt
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above -20°C (-4°F).