missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubidecarenone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 303-98-0 |
| Linear Formula | C59H90O4 |
| MDL Number | MFCD00042919 |
| Synonym | Coenzyme Q10; Q-10; Ubiquinone 50; Ubiquinone-10 |
| Type | Certified Reference Material |
| Quantity | 500 mg |
| Formula Weight | 863.34 |
| Grade | Certified Reference Material |