missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin AG 835 98.0+%, TCI America™

Catalog No. T354025MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T354025MG 25 mg
T3540100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T354025MG Supplier TCI America Supplier No. T354025MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 133550-37-5
Molecular Formula C18H16N2O3
Molecular Weight (g/mol) 308.337
MDL Number MFCD00209855
InChI Key UMGQVUWXNOJOSJ-CDCQXCROSA-N
Synonym tyrphostin b44, + enantiomer, --r-n-, a-methylbenzyl-3,4-dihydroxybenzylidenecyanoacetamide
PubChem CID 23919058
IUPAC Name (Z)-2-cyano-3-(3,4-dihydroxyphenyl)-N-[(1R)-1-phenylethyl]prop-2-enamide
SMILES CC(C1=CC=CC=C1)NC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N

Specifications

Melting Point 146°C
Quantity 25 mg
Formula Weight 308.34
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Tyrphostin AG 835
TSCA No
Recommended Storage Freezer