missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin AG 1478 98.0+%, TCI America™

Catalog No. T294425MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T294425MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T294425MG Supplier TCI America Supplier No. T294425MG
Only null left

Tyrphostin AG 1478 98.0+%, TCI America™, available now.

Specifications

CAS 153436-53-4
Molecular Formula C16H14ClN3O2
Synonym N-(3-Chlorophenyl)-6,7-dimethoxy-4-quinazolinamine
Quantity 25 mg