missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrphostin A1 98.0+%, TCI America™
Specifications
Specifications
| CAS | 2826-26-8 |
| Molecular Formula | C11H8N2O |
| Synonym | Tyrphostin 1, (4-Methoxybenzylidene)malononitrile |
| Quantity | 200 mg |