missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TYRP1 Chimeric Recombinant Human Monoclonal Antibody (TA99)
GREENER_CHOICE

Numéro de catalogue. MA551644
Modifier l'affichage
Click to view available options
Quantité:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
MA551644 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. MA551644 Fournisseur Invitrogen™ Code fournisseur MA551644

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Human Recombinant Monoclonal Antibody

Binds Tyrosinase-related protein-1 (TRP-1), a 70-75k enzyme located in melanocytes, which are specialized cells that produce a pigment called melanin, helping to stabilize tyrosinase, which is the enzyme responsible for the first step in melanin production and determine the shape of melanosomes, which are the structures in melanocytes where melanin is produced.

This gene encodes a melanosomal enzyme that belongs to the tyrosinase family and plays an important role in the melanin biosynthetic pathway. Defects in this gene are the cause of rufous oculocutaneous albinism and oculocutaneous albinism type III.

Spécifications

Antigène TYRP1 Chimeric
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone TA99
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 0.02% ProClin 300
Gène TYRP1
Numéro d’acquisition du gène P17643
Alias du gène 5,6-dihydroxyindole-2-carboxylic acid oxidase; B; b-PROTEIN; brown; Brown locus protein; CAS2; Catalase B; CATB; DHICA oxidase; glycoprotein 75; GP75; iris stromal atrophy; isa; melanogenic enzyme; melanoma antigen gp75; MGC120175; MGC149362; OCA3; RP11-3L8.1; TRP; TRP1; TRP-1; truncated Tyrp1; TRY1; TRY4; TRYP1; tyrosinase related protein 1; tyrosinase-related protein 1; tyrosinase-related protein 1 precursor; tyrosine phosphatase 1, same as B (Brown); TYRP; Tyrp1; Tyrp-1; TYRRP
Symboles de gène TYRP1
Espèces hôtes Human
Immunogène 70-75 kDa pigmentation-associated glycoprotein in human melanoma cell lines.
Méthode de purification Protein A
Quantité 200 μg
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 7306
Espèces cibles Human
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG1 κ
Afficher plus de résultats Afficher moins de résultats