missing translation for 'onlineSavingsMsg'
Learn More

Tyramine hydrochloride, 99%

Catalog No. AC422381000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422381000 100 g Glass bottle
AC422380050 5 g Glass bottle
AC422380250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC422381000 Supplier Thermo Scientific Chemicals Supplier No. 422381000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tyramine hydrochloride, CAS # 60-19-5, is the hydrochloride salt of tyramine, a biogenic amine derived from the amino acid tyrosine. | CAS: 60-19-5 | C8H12ClNO | 173.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Tyramine is a naturally occurring monoamine that is derived from the decarboxylation of the amino acid tyrosine utilized in human and invertebrate nervous systems
  • Tyramine is synthesized and released from neurons and acts as a neuromodulator upon binding to tyramine receptors
  • Tyramine has been found to impact the immune system and cardiovascular system in humans

Applications

  • Tyramine may be used to form tyramide conjugate, which can be used as a signal amplification reagent in immunofluorescence assays
  • Tyramine-hyaluronic acid conjugates can be used to form biodegradable hydrogels, with utility in drug delivery and tissue engineering
  • Tyramine is useful for laboratory studies of neuronal stimulation

Chemical Identifiers

CAS 60-19-5
Molecular Formula C8H12ClNO
Molecular Weight (g/mol) 173.64
MDL Number MFCD00012901
InChI Key RNISDHSYKZAWOK-UHFFFAOYSA-N
Synonym tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
PubChem CID 66449
SMILES [H+].[Cl-].NCCC1=CC=C(O)C=C1

Specifications

Melting Point 271°C to 274°C
Color White
Boiling Point 269°C
Loss on Drying 0.5% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5% (on dry substance) (Argentometry)
Packaging Glass bottle
Linear Formula HOC6H4CH2CH2NH2·HCl
Quantity 100 g
Beilstein 13, 625
Merck Index 15, 10016
Solubility Information (10% in water) Clear colorless to light yellow
IUPAC Name 4-(2-aminoethyl)phenol;hydrochloride
Formula Weight 173.65
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tyramine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 200-462-5

RUO – Research Use Only