missing translation for 'onlineSavingsMsg'
Learn More

Tylosin Tartrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013058
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013058 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013058 Supplier Merck Emd Millipore Supplier No. PHR1508500MG
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Tylosin Tartrate is a tylosin derivative and a medium spectrum antibiotic, which can find applications in veterinary medicine for the treatment of infections caused by most gram-positive bacteria, mycoplasmas, some gram-negative bacteria and chlamydia bacteria. It can also be used as a feed additive in order to improve the weight gain and feed efficiency of cattle, poultry, turkey and swine.

Specifications

CAS 1405-54-5
Linear Formula (C46H77 NO17)x · C4H6O6
Synonym Tylosin (2 R,3 R)-2, 3-dihydroxybutanedioate (salt)
Type Certified Reference Material
Quantity 500 mg
Grade Certified Reference Material