missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TXNL2 Monoclonal Antibody (OTI4C8)
GREENER_CHOICE

Numéro de catalogue. pima527393
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIMA527393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIMA527393 Fournisseur Invitrogen™ Code fournisseur MA527393
Il en reste null

Mouse Monoclonal Antibody

TXNL2 Monoclonal Antibody for Western Blot

This gene encodes a member of the glutaredoxin family. Glutaredoxins are oxidoreductase enzymes that reduce a variety of substrates using glutathione as a cofactor. The encoded protein binds to and modulates the function of protein kinase C theta. The encoded protein may also inhibit apoptosis and play a role in cellular growth, and the expression of this gene may be a marker for cancer. Pseudogenes of this gene are located on the short arm of chromosomes 6 and 9. Alternatively spliced transcript variants have been observed for this gene.

Spécifications

Antigen TXNL2
Applications Western Blot
Classification Monoclonal
Clone OTI4C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GLRX3
Gene Accession No. O76003
Gene Alias bA500G10.4; FLJ11864; Glrx3; GLRX4; glutaredoxin 3; glutaredoxin 4; glutaredoxin-3; GRX3; GRX4; HUSSY-22; Picot; PKC interacting cousin of thioredoxin; PKC-interacting cousin of thioredoxin; PKCq-interacting protein; PKC-theta-interacting protein; testicular tissue protein Li 75; thioredoxin-like 2; thioredoxin-like protein 2; TXNL2; TXNL3
Gene Symbols GLRX3
Host Species Mouse
Immunogen Full length human recombinant protein of GLRX3 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10539
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Afficher plus de résultats Afficher moins de résultats