missing translation for 'onlineSavingsMsg'
Learn More

Tween 85 (=Polyoxyethylene Sorbitan Trioleate), TCI America™

Catalog No. T054725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T054725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T054725G Supplier TCI America Supplier No. T054725G

Chemical Identifiers

CAS 9005-70-3
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7
Molecular Weight (g/mol) NaN
MDL Number MFCD01779641
InChI Key ZGEQUSWSYZZNAI-CLFAGFIQNA-N
Synonym Polyoxyethylene Sorbitan Trioleate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO

Specifications

Color Undesignated
Physical Form Clear Liquid-Cloudy Liquid at 20°C
Melting Point -20°C
Quantity 25 g
Chemical Name or Material Tween 85 (=Polyoxyethylene Sorbitan Trioleate)
EINECSNumber (8)-0055
RTECSNumber WG2934500
TSCA Yes