missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TWEEN 20 (50% Solution)
TWEEN 20 (50% Solution)
Supplier: Invitrogen™ 003005
Description
- May be added to TBS or PBS at low concentrations, to reduce non-specific binding in ELISAs and IHC.
Specifications
| Liquid | |
| 20 mL |
Safety and Handling
Recommended Storage : Store at room temperature.
Shelf life: 18 months from date of manufacture
For Research Use Only. Not for use in diagnostic procedures.