missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Turbidity Stock Solution I, Hydrazine Sulfate, 1%, LabChem™
Supplier: LabChem LC263001
Specifications
| Turbidity Stock Solution I | |
| 10034-93-2 , 7732-18-5 | |
| 99,1 | |
| Liquid | |
| Passes Test | |
| Poly Bottle | |
| 130.12 |
| Hydrazine Sulfate | |
| Colorless | |
| Nitrogen oxides; Gaseous Ammonia | |
| 500 mL | |
| H4N2·H2SO4 | |
| Soluble in water | |
| 1% |