missing translation for 'onlineSavingsMsg'
Learn More

Turbidity Stock Solution I, Hydrazine Sulfate, 1%, LabChem™

Catalog No. p-5388071
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC263007 125 mL
LC263001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC263007 Supplier LabChem Supplier No. LC263007
Only null left

Specifications

Chemical Name or Material Turbidity Stock Solution I
Name Note Hydrazine Sulfate
CAS 10034-93-2, 7732-18-5
Color Colorless
CAS Max % 99, 1
Decomposition Information Nitrogen oxides; Gaseous Ammonia
Physical Form Liquid
Quantity 125 mL
Identification Passes Test
Linear Formula H4N2·H2SO4
Packaging Poly Bottle
Solubility Information Soluble in water
Formula Weight 130.12
Percent Purity 1%
Show More Show Less