missing translation for 'onlineSavingsMsg'
Learn More

Turbidity Stock Solution 1, 1% Hydrazine Sulfate, Ricca Chemical

Catalog No. 88314
Change view
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88314 120 mL
883116 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 88314 Supplier Ricca Chemical Company Supplier No. RI88314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Turbidity Stock Solution 1
Packaging Amber Glass Bottle
CAS 10034-93-2, 7732-18-5
Concentration or Composition Notes 1% Hydrazine Sulfate
CAS Min % 97.0, 0.97
CAS Max % 100.0, 1.01
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 120 mL