missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Turbidity Stock Solution 1, 1% Hydrazine Sulfate, Ricca Chemical
Supplier: Ricca Chemical Company 883116
Specifications
| Turbidity Stock Solution 1 | |
| 10034-93-2 , 7732-18-5 | |
| 1% Hydrazine Sulfate | |
| Liquid | |
| 100°C |
| Amber Glass Bottle | |
| 97.0,0.97 | |
| 100.0,1.01 | |
| Colorless | |
| 500 mL |