missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Turbidity Stock Solution 1, 1% Hydrazine Sulfate, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Turbidity Stock Solution 1 |
| Packaging | Amber Glass Bottle |
| CAS | 10034-93-2, 7732-18-5 |
| Concentration or Composition Notes | 1% Hydrazine Sulfate |
| CAS Min % | 97.0, 0.97 |
| CAS Max % | 100.0, 1.01 |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| Quantity | 120 mL |