Learn More
United Scientific™ Tuning Pipes
These tuning pipes create a long-lasting sound that fills an entire room.
Supplier: United Scientific™ TUNPIP256
Alert:
Prop 65
Description
Compared to traditional tuning forks, they are much louder and have a more penetrating tone. Made from an aluminum alloy, each pipe is tuned to a specific frequency. The tuning pipe is supported by a wooden handle and string, allowing it to vibrate freely without any dampening from being held. The frequency is laser etched onto each pipe for easy reference. For best results, a rubber hammer is included to activate the pipe.
- Carefully tuned to each frequency
- Made from high quality, heat-treated aluminum
- Louder than traditional tuning forks, with a long-lasting sound
- Available in 256 Hz., 384 Hz., and 528 Hz., and as a set of three
Chemical Identifiers
| Tuning Pipe |
Specifications
| Tuning Pipe | |
| 256 Hz |