Learn More
United Scientific™ Tuning Fork
Tuning forks are available in many different frequencies.
Supplier:  United Scientific™ TFK256
 Afficher plus de versions de ce produit
Numéro de catalogue. S16030
Il en reste null
Description
Description
  • Made of aluminum alloy.
  • The frequency and scale letter is stamped on each fork.
  • Available in a wide variety of frequencies.
Spécifications
Identifiants chimiques
Tuning Fork
Spécifications
Tuning Fork
256 Hz
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS