missing translation for 'onlineSavingsMsg'
Learn More

Tungsten sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis)

Catalog No. AA41639KS
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41639KS 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41639KS Supplier Thermo Scientific Chemicals Supplier No. 041639KS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-33-7 | W | 183.84 g/mol

Tungsten was used extensively for the filaments of old-style incandescent light bulbs, its alloys are used in many high-temperature applications, such as arc-welding electrodes and heating elements in high-temperature furnaces. Some bacteria use tungsten in an enzyme to reduce carboxylic acids to aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Quantity 1 Ea.
Solubility Information Insoluble in water.
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Tungsten sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
EINECSNumber 231-143-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only