missing translation for 'onlineSavingsMsg'
Learn More

Tungsten powder, -325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog No. AA39749A3
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39749A3 2 kg
AA3974922 100 g
AA3974936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA39749A3 Supplier Thermo Scientific Chemicals Supplier No. 039749A3
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Appearance Light Gray powder
Quantity 2 kg
Mesh Size -325 Mesh
Form Powder
Assay Percent Range 99.9% (Metals basis)
Chemical Name or Material Tungsten
Hazard Category H228-H500
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 231-143-9
RTECSNumber YO7175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only