missing translation for 'onlineSavingsMsg'
Learn More

Tungsten ICP Standard, 10,000 ppm W in H2O/tr NH4OH, Ricca Chemical

Supplier:  Ricca Chemical Company PW10KW500

Packaging Natural Poly Bottle

Catalog No. PW10KW500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

11120-25-5, 1336-21-6, 7732-18-5

Specifications

Tungsten ICP Standard
11120-25-5 , 1336-21-6 , 7732-18-5
100.0, 1.37, 0.5
1mL = 10mg W (10, 000ppm W)(NH4)2WO4 in H2O / tr NH4OH
Natural Poly Bottle
96.0, 1.32, 0.48
Odorless
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 months

SDS
Documents

Documents