Learn More
Tungsten ICP Standard, 1000 ppm W in H2O/tr NH4OH, Ricca Chemical
Single Element ICP Standard
Supplier:  Ricca Chemical Company RPW1KW00100N
Packaging Natural LDPE Bottle
Catalog No. RPW1KW00100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Specifications
Chemical Identifiers
11120-25-5, 1336-21-6, 7732-18-5
Specifications
Tungsten ICP Standard, 1000 ppm W in H2O/tr NH4OH
11120-25-5 , 1336-21-6 , 7732-18-5
99.74, 0.14, 0.13
Miscible in water
Natural LDPE Bottle
99.74, 0.14, 0.13
1
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
ShelfLife : 18 months
Recommended Storage : Ambient