missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten ICP Standard, 1000 ppm W in H2O/tr NH4OH, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 97.0, 0.49, 0.14 |
| CAS Max % | 100.0, 0.51, 0.14 |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| Type | Standard |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | 1mL = 1mg W (1, 000ppm W)(NH4)2WO4 in H2O / tr NH4OH |
| Chemical Name or Material | Tungsten ICP Standard |
Safety and Handling
| ShelfLife | 18 months |