missing translation for 'onlineSavingsMsg'
Learn More

Tungsten AA Standard, 1000 ppm W in H2O, Ricca Chemical

Supplier:  Ricca Chemical Company AW1KW100

Packaging Natural LDPE Bottle

Catalog No. AW1KW100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

11120-25-5, 1336-21-6, 7732-18-5

Specifications

Tungsten AA Standard
11120-25-5 , 1336-21-6 , 7732-18-5
100.0, 0.51, 0.14
Natural LDPE Bottle
97.0, 0.49, 0.13
1, 000ppm in H2O / 0.5% NH4OH
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 months

SDS
Documents

Documents