missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Tulobuterol Hydrochloride, 98%
beta-2-adrenergic receptor agonist | CAS: 56776-01-3 | C12H18ClNO·HCl
$298.88 - $774.92
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ6144803
|
Thermo Scientific Chemicals
J6144803 |
1 g |
Each for $298.88
|
|
|||||
|
AAJ6144806
|
Thermo Scientific Chemicals
J6144806 |
5 g |
Each for $774.92
|
|
|||||
Description
beta-2-adrenergic receptor agonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 56776-01-3 | |
| C12H18ClNO·HCl |
Specifications
| White | |
| beta-2-adrenergic receptor agonist | |
| C12H18ClNO·HCl | |
| Hygroscopic | |
| Soluble in methanol or water | |
| Powder |
| 2°C to 8°C | |
| 56776-01-3 | |
| 264.19 | |
| 159°C to 163°C | |
| Signal Transduction Reagent | |
| 98% |
Safety and Handling
Recommended Storage : Keep cold
RUO – Research Use Only