missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tulobuterol Hydrochloride, 98%

Catalog No. AAJ6144806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6144806 5 g
AAJ6144803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6144806 Supplier Thermo Scientific Chemicals Supplier No. J6144806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

beta-2-adrenergic receptor agonist | CAS: 56776-01-3 | C12H18ClNO·HCl

beta-2-adrenergic receptor agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56776-01-3
Molecular Formula C12H18ClNO·HCl

Specifications

Quantity 5 g
Description White
Content And Storage 2°C to 8°C
For Use With (Application) beta-2-adrenergic receptor agonist
Formula Weight 264.19
Sensitivity Hygroscopic
Melting Point 159°C to 163°C
Solubility Soluble in methanol or water
Product Type Signal Transduction Reagent
Form Powder
Assay 98%
Show More Show Less

RUO – Research Use Only