CAS RN 56776-01-3
IUPAC Name:
hydrogen 2-(tert-butylamino)-1-(2-chlorophenyl)ethan-1-ol chloride
Synonyms:
tulobuterol hydrochloride
Molecular Weight (g/mol):
264.19
Molecular Formula:
C12H19Cl2NO
InChi Key:
RSLNRVYIRDVHLY-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CC(C)(C)NCC(O)C1=CC=CC=C1Cl
1
–
3
of
3
results
1
Thermo Scientific Chemicals Tulobuterol Hydrochloride, 98%
CAS: 56776-01-3 Molecular Formula: C12H18ClNO·HCl
2
VETRANAL™ Tulobuterol hydrochloride, ≥98% (HPLC), MilliporeSigma™ Supelco™
Tulobuterol hydrochloride is a sympathomimetic amine, possessing bronchodilator activity.
3
Tulobuterol Hydrochloride 98.0+%, TCI America™
CAS: 56776-01-3 Molecular Formula: C12H19Cl2NO Molecular Weight (g/mol): 264.19 MDL Number: MFCD00214398 InChI Key: RSLNRVYIRDVHLY-UHFFFAOYSA-N Synonym: 2-tert-Butylamino-1-(2-chlorophenyl)ethanol Hydrochloride PubChem CID: 5702285 ChEBI: CHEBI:32272 IUPAC Name: 2-(tert-butylamino)-1-(2-chlorophenyl)ethanol;hydrochloride SMILES: CC(C)(C)NCC(C1=CC=CC=C1Cl)O.Cl