missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ FBV Value Manual Defrost Freezer

Catalog No. FBV05FPSA
Change view
Click to view available options
Capacity (English):
5 cu. ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
FBV05FPSA 5 cu. ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FBV05FPSA Supplier Fisherbrand™ Supplier No. FBV05FPSA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The FBV Value Manual Defrost Laboratory Freezer offers an economical, simple, and environmentally friendly solution for safe and reliable general-purpose storage.

  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
  • Adjustable temperature control
  • Easy to clean, durable finish

Specifications

Type Undercounter Freezer
Capacity (English) 5 cu. ft.
Capacity (Metric) 141.5 L
Temperature Range -12°C to -24°C
Door Style Solid Swing
No. of Doors 1
Insulation Polyurethane Foam
Interior ABS Plastic
Exterior Finish Enameled Steel
Shelves 2 Fixed
Door Shelves 3
Style Undercounter
Casters 2 adjustable front feet
Color White
Control Type Mechanical Dial
For Use With (Application) Indoor general storage of non-critical samples and reagents
Typical Lifetime 10 Years
Warranty 13 Months
Defrost Manual
Min. Clearance 0.5 in. on the top and sides, 4 in. on the rear
Dimensions (DxWxH) Exterior 25.8 x 21.4 x 33.6 in. (65.5 x 54.3 x 85.3 cm)
Dimensions (D x W x H) Interior 19.0 x 18.6 x 29.5 in. (48.2 x 47.3 x 74.9 cm)
Monitoring Options Available Separately
Access Security None
Amperage 1.4 A
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115 V, 60 Hz
Frequency 60 Hz
Energy Usage 1.3 kWh/Day
Compressor Hermetic
Refrigerant R600a Hydrocarbon Refrigerant
Shipping Weight (English) 95 lb.
Shipping Weight (Metric) 43.1 kg
Show More Show Less