missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSV Value Convertible Freezer/Refrigerator

Catalog No. TSV20RPSA
Change view
Click to view available options
Type:
General Purpose Freezer (Convertible to Refrigerator)
General Purpose Refrigerator (Convertible to Freezer)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
TSV20RPSA General Purpose Refrigerator (Convertible to Freezer)
TSV20FPSA General Purpose Freezer (Convertible to Refrigerator)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. TSV20RPSA Supplier Thermo Scientific™ Supplier No. TSV20RPSA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Flexible and economical laboratory storage.

Thermo Scientific TSV Value freezers and refrigerators offer economical and simple sample storage while also offering increased flexibility of operating as either a refrigerator or a freezer with the push of a button.

The TSV20FSPSA ships as an auto defrost freezer and the TSV20RPSA ships as a refrigerator, but each easily converts to a refrigerator or an auto defrost freezer respectively.

  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
  • Adjustable temperature control
  • Easy to clean, durable finish

Specifications

Type General Purpose Refrigerator (Convertible to Freezer)
Capacity (English) 21 cu. ft.
Capacity (Metric) 594 L
Temperature Range 1°C to 12°C or -12°C to -24°C
Temperature (Setpoint) Adjustable
Refrigerant R600a
Door Style Hinged
No. of Doors 1 (1 compartment)
Hinge Side Right
Shelves 3 Adjustable
Door Shelves 4 fixed shelves
Cabinet Material Cold-rolled Steel
Exterior Finish White-enameled Steel
Insulation Cyclopentane
Interior ABS Plastic
Certifications/Compliance cULus
Style Upright
Casters 2 adjustable leveling feet
Color White
Control Type Electronic control
For Use With (Application) Indoor general storage of non-critical samples and reagents
Typical Lifetime 10 years
Warranty 13 months parts and labor
Defrost Automatic
Lighting Interior
Min. Clearance 4 in. on sides, 4 in. at back, 11 in. on top
Monitoring Options Remote Alarm Contacts
Access Security None
Operating Modes 2 - refrigerator or freezer
Compressor Bottom-mounted
Dimensions (D x W x H) Interior 22.5 x 26.5 x 69 in (57.15 x 67.3 x 175.26 cm)
Dimensions (L x W x H) Exterior 28.94 x 32.76 x 76.77 in. (73.5 x 83.2 x 195 cm)
Amperage 2.2A
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115 V/60 Hz
Frequency 50 Hz
Shipping Weight (English) 244 lb.
Shipping Weight (Metric) 110.6 kg
Show More Show Less