missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSNAX Monoclonal Antibody (K52014_14D2), Invitrogen™

Numéro de catalogue. MA555853
Modifier l'affichage
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MA555853 50 μL
MA556233 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MA555853 Fournisseur Invitrogen™ Code fournisseur MA555853
Il en reste null

Mouse Monoclonal Antibody

Sequence of this protein is as follows: MSNKEGSGGF RKRKHDNFPH NQRREGKDVN SSSPVMLAFK SFQQELDARH DKYERLVKLS RDITVESKRT IFLLHRITSA PDMEDILTES EIKLDGVRQK IFQVAQELSG EDMHQFHRAI TTGLQEYVEA VSFQHFIKTR SLISMDEINK QLIFTTEDNG KENKTPSSDA QDKQFGTWRL RVTPVDYLLG VADLTGELMR MCINSVGNGD IDTPFEVSQF LRQVYDGFSF IGNTGPYEVS KKLYTLKQSL AKVENACYAL KVRGSEIPKH MLADVFSVKT EMIDQEEGIS

This gene encodes a protein which specifically interacts with translin, a DNA-binding protein that binds consensus sequences at breakpoint junctions of chromosomal translocations. The encoded protein contains bipartite nuclear targeting sequences that may provide nuclear transport for translin, which lacks any nuclear targeting motifs.

Spécifications

Antigen TSNAX
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone K52014_14D2
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene TSNAX
Gene Accession No. Q99598
Gene Alias R74833; translin associated factor X; translin-associated factor X; Translin-associated protein X; translin-like protein; Trax; TSNAX
Gene Symbols TSNAX
Host Species Mouse
Immunogen Recombinant human TSNAX (immunogen range is 1-290).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7257
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Afficher plus de résultats Afficher moins de résultats